Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VAT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595877
Description
Immunogen sequence: GVGMAAVQLC RTVENVTVFG TASASKHEAL KENGVTHPID YHTTDYVDEI KKISPKGVDI VMDPLGGSDT AKGYNLLKPM GKVVTYGMAN LLTGPKRNLM A.
Synaptic vesicles are responsible for regulating the storage and release of neurotransmitters in the nerve terminal. The protein encoded by this gene is an abundant integral membrane protein of cholinergic synaptic vesicles and is thought to be involved in vesicular transport. It belongs to the quinone oxidoreductase subfamily of zinc-containing alcohol dehydrogenase proteins.
Specifications
VAT1 | |
Polyclonal | |
Unconjugated | |
VAT1 | |
FLJ20230; membrane protein of cholinergic synaptic vesicles; mitofusin-binding protein; Protein MIB; si:dz163l24.2; solute carrier family 18 (vesicular monoamine), member 1; synaptic vesicle membrane protein VAT-1 homolog; VAT1; Vat-1; vat1 protein; VATI; vesicle amine transport 1; vesicle amine transport protein 1 homolog (T californica); vesicle amine transport protein 1 homolog (T. californica) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10493, 26949, 287721 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.13 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q3MIE4, Q62465, Q99536 | |
VAT1 | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human VAT1 (NP_0063642). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction