Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAT1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501733684 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3684 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3684 Supplier Proteintech Group Inc Supplier No. 220161AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VAT1 gene ontology annotations related to this gene include negative regulation of mitochondrial fusion.

Specifications

Antigen VAT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene VAT1
Gene Accession No. Q3MIE4, Q99536
Gene Alias VAT1, VATI
Gene Symbols Vat1
Host Species Rabbit
Immunogen VAT1 Fusion Protein Ag16652
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10493, 287721
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.