Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAT1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598934
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598934 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598934 Supplier Invitrogen™ Supplier No. PA598934
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VAT1L Polyclonal Antibody for Western Blot, IHC (P), ELISA

VAT1L is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAT1L
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene VAT1L
Gene Accession No. Q9HCJ6
Gene Alias 9430073I07; AI427515; Kiaa1576; mKIAA1576; Synaptic vesicle membrane protein VAT-1 homolog-like; Vat1l; vesicle amine transport 1-like; vesicle amine transport protein 1 homolog (T. californica)-like; vesicle amine transport protein 1 homolog-like (T. californica)
Gene Symbols VAT1L
Host Species Rabbit
Immunogen Recombinant Human Synaptic vesicle membrane protein VAT-1 homolog-like protein (1-419AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57687
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.