Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vav (pan) Antibody (SR2313), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32265850UL
Description
Vav (pan) Monoclonal antibody specifically detects Vav (pan) in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
Vav (pan) | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
A synthesized peptide derived from human Vav (pan) (Uniprot #: P15498, P52735, Q9UKW4) | |
50 μg | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
SR2313 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Rabbit | |
Affinity purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction