Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAV2 Antibody (3F5D3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | VAV2 |
---|---|
Clone | 3F5D3 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry |
Description
VAV2 Monoclonal specifically detects VAV2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
VAV2 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Signal Transduction | |
PBS with 0.05% Sodium Azide | |
7410 | |
Purified recombinant fragment of human VAV2 (AA: 552-868) expressed in E. Coli. | |
Primary | |
101.3 kDa |
3F5D3 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
guanine nucleotide exchange factor VAV2, oncogene VAV2, Protein vav-2, vav 2 guanine nucleotide exchange factor, vav 2 oncogene, VAV-2 | |
VAV2 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title