Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAV2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323273
Description
VAV2 Polyclonal antibody specifically detects VAV2 in Human, Mouse, Rat, Chicken samples. It is validated for Western BlotSpecifications
VAV2 | |
Polyclonal | |
Western Blot 1:1000 | |
guanine nucleotide exchange factor VAV2, oncogene VAV2, Protein vav-2, vav 2 guanine nucleotide exchange factor, vav 2 oncogene, VAV-2 | |
A synthetic peptide (coupled to KLH) corresponding to amino acid residues in the Dbl homology domain of human Vav2. This sequence has high homology with similar regions in rat and mouse Vav2, and has low homology to other Vav family members. [UniProt# P52735] | |
0.1 mL | |
Signal Transduction | |
7410 | |
Store at -20°C. | |
IgG |
Western Blot | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction