Learn More
Invitrogen™ VAV2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585696
Description
Keep as concentrated solution. Predicted reactivity: Mouse (95%), Rat (95%), Chicken (91%), Rhesus Monkey (100%). Positive Control: 293T, A431, HeLa, HepG2, mouse olfactory bulb. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
VAV2 is the second member of the VAV guanine nucleotide exchange factor family of oncogenes. Unlike VAV1, which is expressed exclusively in hematopoietic cells, VAV2 transcripts were found in most tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
VAV2 | |
Polyclonal | |
Unconjugated | |
Vav2 | |
2810040F13Rik; AI847175; Guanine nucleotide exchange factor VAV2; Oncogene VAV2; vav 2 guanine nucleotide exchange factor; vav 2 oncogene; vav guanine nucleotide exchange factor 2; VAV2; VAV-2; Vav2 oncogene | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22325, 7410 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
0.96 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P52735, Q60992 | |
Vav2 | |
Recombinant protein encompassing a sequence within the C-terminus region of human VAV2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.