Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAV2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987090
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7090 Supplier Bioss Supplier No. BS6160R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VAV2 is the second member of the VAV guanine nucleotide exchange factor family of oncogenes. Unlike VAV1, which is expressed exclusively in hematopoietic cells, VAV2 transcripts were found in most tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen VAV2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Vav2
Gene Accession No. P52735
Gene Alias 2810040F13Rik; AI847175; Guanine nucleotide exchange factor VAV2; Oncogene VAV2; vav 2 guanine nucleotide exchange factor; vav 2 oncogene; vav guanine nucleotide exchange factor 2; VAV2; VAV-2; Vav2 oncogene
Gene Symbols Vav2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human VAV2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7410
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.