Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAV3 Antibody (SR2314), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32216550UL
Description
VAV3 Monoclonal antibody specifically detects VAV3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
VAV3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
SR2314 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:40, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
FLJ40431, guanine nucleotide exchange factor VAV3, vav 3 guanine nucleotide exchange factor, vav 3 oncogene, VAV-3 | |
A synthesized peptide derived from human VAV3 (Uniprot #: Q9UKW4) | |
50 μg | |
Phospho Specific, Signal Transduction | |
10451 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction