Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564931
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564931 Supplier Invitrogen™ Supplier No. PA564931
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRAESGGGGGR CGDRSGAGDL RADGGGHSPT EVAGTSASSP AGSRESGADS DGQPGPGEAD HCRRILVRDA KGTIREIVLP KGLDLDRPKR T Highest antigen sequence identity to the following orthologs - mouse 79%, rat 78%.

This gene encodes a homeobox protein and is almost exclusively expressed in the ventral portion of the retina during development. In mouse studies, this gene was found to be required for the correct formation of the optic fissure and other aspects of retinal development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAX2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VAX2
Gene Accession No. Q9UIW0
Gene Alias DRES93; VAX2; Ventral anterior homeobox 2; ventral anterior homeobox containing gene 2; ventral retina homeodomain protein; Vex
Gene Symbols VAX2
Host Species Rabbit
Immunogen Recombinant Human VAX2. Recombinant protein control fragment (Product #RP-105718).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25806
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.