Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598238
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598238 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598238 Supplier Invitrogen™ Supplier No. PA598238
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VAX2 Polyclonal Antibody for Western Blot, ELISA

This gene encodes a homeobox protein and is almost exclusively expressed in the ventral portion of the retina during development. In mouse studies, this gene was found to be required for the correct formation of the optic fissure and other aspects of retinal development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAX2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene VAX2
Gene Accession No. Q9UIW0
Gene Alias DRES93; VAX2; Ventral anterior homeobox 2; ventral anterior homeobox containing gene 2; ventral retina homeodomain protein; Vex
Gene Symbols VAX2
Host Species Rabbit
Immunogen Recombinant Human Ventral anterior homeobox 2protein (1-74AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25806
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.