Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VBP1 Antibody (OTI2E6), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274836R
Description
VBP1 Monoclonal specifically detects VBP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
VBP1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
VBP1 | |
Full length recombinant protein of human VBP1 (NP_003363) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2E6 | |
Western Blot, Immunohistochemistry | |
HIBBJ46, PFD3, PFDN3VHL-binding protein 1, prefoldin 3, prefoldin subunit 3, VBP-1, VHL binding protein-1, von Hippel-Lindau binding protein 1, Von Hippel-Lindau-binding protein 1 | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
7411.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction