Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VBP1 Monoclonal Antibody (OTI1G3), TrueMAB™, OriGene
SDP

Catalog No. 502540039
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0039 30 μL
50-167-5501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0039 Supplier OriGene Supplier No. TA504912S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VBP1 Monoclonal antibody specifically detects VBP1 in Canine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen VBP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1G3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Vbp1
Gene Accession No. P61758, P61759
Gene Alias EGK_21124; HIBBJ46; LOC703257; PFD3; Pfdn3; phosphatidylinositol-binding clathrin assembly protein; prefoldin 3; prefoldin subunit 3; prefoldin subunit 3; phosphatidylinositol-binding clathrin assembly protein; RP13-228J13.4; VBP1; VBP-1; VHL binding protein 1; VHL binding protein-1; VHL-binding protein 1; von Hippel-Lindau binding protein 1; von Hippel-Lindau-binding protein 1
Host Species Mouse
Immunogen Full length human recombinant protein of human VBP1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22327, 481089, 681825, 7411
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.