Learn More
VBP1 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI2B12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504904

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by VBP1 interacts with the Von Hippel-Lindau protein to form an intracellular complex. The encoded protein functions as a chaperone protein, and may play a role in the transport of the Von Hippel-Lindau protein from the perinuclear granules to the nucleus or cytoplasm. Alternative splicing and the use of alternate transcription start sites results in multiple transcript variants encoding different protein isoforms.Specifications
VBP1 | |
Monoclonal | |
Unconjugated | |
VBP1 | |
HIBBJ46, PFD3, PFDN3, VBP-1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
22327, 481089, 681825, 7411 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2B12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P61758, P61759 | |
VBP1 | |
Full length human recombit protein of human VBP1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.