Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VBta 17a TCR Mouse, anti-Mouse, R718, Clone: KJ23, BD OptiBuild™

Catalog No. BDB751964 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751964 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751964 Supplier BD Biosciences Supplier No. 751964
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The KJ23 monoclonal antibody specifically recognizes Vβ 17[a] T-cell Receptor (TCR) of mice having the a haplotype (eg,C57L, SJL, SWR) of the Tcrb gene complex. Strains having the b (eg, A, AKR, BALB/c, CBA,C3H/He, C57BL, C58, DBA/1, DBA/2) Tcrb haplotype do not express functional Vβ 17 TCR, and the Tcrb-V17 gene locus is deleted in mice having the c (eg, RIII) haplotype. Vβ 17[a] TCR-bearing Tlymphocytes are clonally eliminated in mice expressing I-E (eg, C57BR). KJ23 antibody also recognizes two phenotypic variants of the Vβ 17[a] TCR: Vβ 17[a2] expressed in a variety of wild-derivedmouse strains and Vβ 17[a(cz)] expressed in Mtv-free CZ mice. The effects of Mtv-encoded superantigens upon Vβ 17[a] TCR-bearing T cells has been reviewed. Plate-bound KJ23 antibody activates Vβ 17[a] TCR-bearing T cells, and injection of the antibody can deplete Vβ 17[a]-bearing Tcells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen VBta 17a TCR
Applications Flow Cytometry
Classification Monoclonal
Clone KJ23
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.