Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VCAM-1/CD106 Antibody (1.G11B1), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128407
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Human, Porcine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
VCAM-1/CD106 | |
Monoclonal | |
PE | |
CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human CD106/VCAM-1 | |
Store at 4C. Do not freeze. | |
IgG1 κ |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
1.G11B1 | |
Western Blot 1:100-1:2000, Flow Cytometry 10 uL/10^6 cells, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
VCAM1 | |
The immunogen is a peptide made to the amino acid region VCAM1 | |
1 mL | |
Mesenchymal Stem Cell Markers, Stem Cell Markers | |
7412 | |
Human, Pig | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction