Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VCAM-1/CD106 Antibody (130), Alexa Fluor™ 647

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290458AF647
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Mouse samples. It is validated for ELISA.Specifications
VCAM-1/CD106 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human VCAM1 Human ICAM1 Human ICAM2 Human ICAM3 Human SELL Human SELP | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
130 | |
ELISA | |
CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse VCAM-1/CD106 (NP_035823.3; Met1-Glu698). | |
0.1 mL | |
Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
7412 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction