Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VCAM-1/CD106 Antibody (6G9), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147491AF750
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
VCAM-1/CD106 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
VCAM1 | |
Purified recombinant fragment of human VCAM-1 expressed in E. coli. [Uniprot: P19320] | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoblot | |
6G9 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting | |
CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
Mouse | |
Ammonium sulfate precipitation | |
Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
7412.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction