Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VCAM-1/CD106 Antibody (6G9) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281005
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Human, Mouse samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
VCAM-1/CD106 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1:500-1:2000, Simple Western 1:1000, ELISA : 1:10000, Immunohistochemistry : 1:200-1:1000, Immunoprecipitation reported in scientific literature (PMID 28569748), Immunohistochemistry-Paraffin : 1:200-1:1000, Immunoblotting reported in scientific literature (PMID 28569748) | |
CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot | |
6G9 | |
Unconjugated | |
PBS | |
VCAM1 | |
Purified recombinant fragment of human VCAM-1 expressed in E. coli. [Uniprot: P19320] | |
0.1 mL | |
Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
7412 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction