Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VCAM-1/CD106 Antibody (6G9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications
$206.50 - $499.50
Specifications
Antigen | VCAM-1/CD106 |
---|---|
Clone | 6G9 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:2000, Simple Western 1:1000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunoprecipitation reported in scientific literature (PMID 28569748), Immunohistochemistry-Paraffin 1:200-1:1000, Immunoblotting reported in scientific literature (PMID 28569748) |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP147491SS
![]() |
Novus Biologicals
NBP147491SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP147491
![]() |
Novus Biologicals
NBP147491 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Human, Mouse samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
VCAM-1/CD106 | |
1.0 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
P19320 | |
7412 | |
Purified recombinant fragment of human VCAM-1 expressed in E. coli. [Uniprot: P19320] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
6G9 | |
Western Blot 1:500-1:2000, Simple Western 1:1000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunoprecipitation reported in scientific literature (PMID 28569748), Immunohistochemistry-Paraffin 1:200-1:1000, Immunoblotting reported in scientific literature (PMID 28569748) | |
Monoclonal | |
Precipitation | |
RUO | |
Human, Mouse | |
CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
VCAM1 | |
IgG1 | |
Ammonium sulfate precipitation | |
81 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title