Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VCAM-1 (CD106) Monoclonal Antibody (6C7.1)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA549138
Description
VCAM-1 (CD106) Monoclonal Antibody for ICC/IF, IHC (F), Flow, IP, Functional Assay
VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.
Specifications
VCAM-1 (CD106) | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% BSA and no preservative | |
P29533 | |
VCAM1 | |
Mouse endothelial cells. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation, Immunocytochemistry | |
6C7.1 | |
Unconjugated | |
VCAM1 | |
adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B | |
Rat | |
Protein G | |
RUO | |
22329 | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction