Learn More
Invitrogen™ VCAM-1 Monoclonal Antibody (6G9)

Description
MA5-15636 targets VCAM1 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15636 immunogen is purified recombinant fragment of human VCAM1 expressed in E. Coli. MA5-15636 detects VCAM1 which has a predicted molecular weight of approximately 81kDa.
Specifications
Specifications
| Antigen | VCAM-1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 6G9 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | VCAM1 |
| Gene Accession No. | P19320 |
| Gene Alias | adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.