Learn More
Invitrogen™ VCAM-1 Monoclonal Antibody (STA)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA110065
Description
This antibody recognizes an extracellular epitope of CD106 antigen (VCAM-1), a 100-110 kDa type I membrane protein of the immunoglobulin superfamily, a crucial mediator of leukocyte adhesion, and a costimulation molecule. Immunohistochemistry (Frozen): acetone fixation.
VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.
Specifications
VCAM-1 | |
Monoclonal | |
1 mg/mL | |
PBS with 15 mM sodium azide; pH 7.4 | |
P19320 | |
VCAM1 | |
Human DS6 T cell line. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
STA | |
Unconjugated | |
VCAM1 | |
adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B | |
Mouse | |
Protein A | |
RUO | |
7412 | |
4°C, do not freeze | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.