Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VCAM1 Monoclonal Antibody (OTI4F1), TrueMAB™, OriGene
SDP

Catalog No. 501673423
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-3423 100 μL
50-254-1602 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-3423 Supplier OriGene Supplier No. TA502361
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VCAM-1 (vascular cell adhesion protein 1, CD106, INCAM-110) is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. VCAM-1 has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases. VCAM-1 is distributed across non-leukocyte and leukocyte cells. VCAM-1 expression is reportedly correlated with adhesion of melanoma, Burkitt lymphoma, osteosarcoma, and kidney carcinoma but not colon carcinoma cells to endothelial cells which may determine the site of tumor metastasis. VCAM-1 is a member of the Ig superfamily of adhesion molecules, is expressed at high levels on cytokine stimulated vascular endothelial cells, and at minimal levels on unstimulated endothelial cells. VCAM-1 is also present on follicular and inter-follicular dendritic cells of lymph nodes, myoblasts, and some macrophages. VCAM-1 serves as a ligand for leukocyte integrin alpha-beta (VLA-4 or CD49d/CD29) and mediates cell adhesion of leukocytes to activated endothelium. VCAM-1 plays a role in tumor metastasis, and may also be involved in the development of atherosclerosis and rheumatoid arthritis. Two alternatively spliced transcripts encoding different isoforms of VCAM-1 have been described.

Specifications

Antigen VCAM1
Applications Western Blot
Classification Monoclonal
Clone OTI4F1
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene VCAM1
Gene Accession No. P19320
Gene Alias CD106, INCAM-100
Gene Symbols VCAM1
Host Species Mouse
Immunogen Full length human recombit protein of human VCAM1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7412
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.