Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VCAM1 Mouse anti-Canine, Human, Clone: OTI3H10, lyophilized, TrueMAB™
SDP

Catalog No. 501661300
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-1300 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-1300 Supplier OriGene Supplier No. CF502391
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

VCAM-1 (vascular cell adhesion protein 1, CD106, INCAM-110) is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. VCAM-1 has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases. VCAM-1 is distributed across non-leukocyte and leukocyte cells. VCAM-1 expression is reportedly correlated with adhesion of melanoma, Burkitt lymphoma, osteosarcoma, and kidney carcinoma but not colon carcinoma cells to endothelial cells which may determine the site of tumor metastasis. VCAM-1 is a member of the Ig superfamily of adhesion molecules, is expressed at high levels on cytokine stimulated vascular endothelial cells, and at minimal levels on unstimulated endothelial cells. VCAM-1 is also present on follicular and inter-follicular dendritic cells of lymph nodes, myoblasts, and some macrophages. VCAM-1 serves as a ligand for leukocyte integrin alpha-beta (VLA-4 or CD49d/CD29) and mediates cell adhesion of leukocytes to activated endothelium. VCAM-1 plays a role in tumor metastasis, and may also be involved in the development of atherosclerosis and rheumatoid arthritis. Two alternatively spliced transcripts encoding different isoforms of VCAM-1 have been described.

Specifications

Antigen VCAM1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3H10
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene VCAM1
Gene Accession No. P19320, Q28260
Gene Alias CD106, INCAM-100
Gene Symbols VCAM1
Host Species Mouse
Immunogen Full length human recombit protein of human VCAM1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 403982, 7412
Target Species Canine, Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.