Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vcan Mouse anti-Human, Mouse, Rat, FITC, Clone:S351-23, Abnova™

Catalog No. 89333331 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-331 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-331 Supplier Abnova Corporation Supplier No. MAB16851
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant mouse Vcan.

Specifications

Antigen Vcan
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone S351-23
Conjugate FITC
Description versican
Dilution Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Vcan
Gene Alias 5430420N07Rik, 9430051N09, Cspg2, DPEAAE, NG2, PG-M, PG-M(V0), PG-M(V1), hdf
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 362-585 at glycosaminoglycan alpha domain of mouse Vcan core protein.
Quantity 100 μg
Gene ID (Entrez) 13003
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.