Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vcan, Mouse anti-Mouse, ALP, Clone: S351-23, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB18513
Specifications
Vcan | |
Monoclonal | |
Alkaline Phosphatase (AP) | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
5430420N07Rik/9430051N09/Cspg2/DPEAAE/NG2/PG-M/PG-M(V0)/PG-M(V1)/hdf | |
Mouse | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
S351-23 | |
Immunocytochemistry (1:100) Immunofluorescence (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
versican | |
Vcan | |
Recombinant protein corresponding to amino acids 362-585 at glycosaminoglycan alpha domain of mouse Vcan core protein. | |
RUO | |
13003 | |
Store at -20°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction