Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VCATH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144425
Description
VCATH Polyclonal Antibody for Western Blot, ELISA
Cysteine protease that plays an essential role in host liquefaction to facilitate horizontal transmission of the virus. Accumulates within infected cells as an inactive proenzyme (proV-CATH), which is activated by proteolytic cleavage upon cell death.
Specifications
VCATH | |
Polyclonal | |
Unconjugated | |
VCATH | |
VCATH; Viral cathepsin | |
Rabbit | |
Protein G | |
RUO | |
1403960 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2.799 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P25783 | |
VCATH | |
Recombinant Autographa californica nuclear polyhedrosis virus Viral cathepsin (VCATH) (113-323aa). | |
100 μL | |
Primary | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction