Learn More
VCIP135 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302933AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Acts as a deubiquitinating enzyme. Necessary for VCP-mediated reassembly of Golgi stacks after mitosis. May play a role in VCP-mediated formation of transitional endoplasmic reticulum (tER). Mediates dissociation of the ternary complex containing STX5A, NSFL1C and VCP. Hydrolyzes ′Lys-11′- and ′Lys-48′-linked polyubiquitin chains.Specifications
VCIP135 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q96JH7 | |
VCPIP1 | |
Between 1100 and 1150 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
VCPIP1 | |
DUBA3, VCIP135, deubiquitinating protein VCIP135, valosin-containing protein (p97)/p47 complex-interacting protein p135, valosin-containing protein p97/p47 complex-interacting protein 1, valosin-containing protein p97/p47 complex-interacting protein p135 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80124 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.