Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VCPIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554933
Description
Immunogen sequence: LLCGALSELH VPPEWLAPGG KLYNLAKSTH GQLRTDKNYS FPLNNLVCSY DSVKDVLVPD YGMSNLTACN WCHGTSVRKV RGDGSIVYLD G Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 95%.
Acts as a deubiquitinating enzyme. Necessary for VCP-mediated reassembly of Golgi stacks after mitosis. May play a role in VCP-mediated formation of transitional endoplasmic reticulum (tER). Mediates dissociation of the ternary complex containing STX5A, NSFL1C and VCP. Hydrolyzes 'Lys-11'- and 'Lys-48'-linked polyubiquitin chains.
Specifications
VCPIP1 | |
Polyclonal | |
Unconjugated | |
VCPIP1 | |
4932442A08; 5730421J18Rik; 5730538E15Rik; Deubiquitinating protein VCIP135; DUBA3; KIAA1850; mKIAA1850; valosin containing protein (p97)/p47 complex interacting protein 1; valosin containing protein interacting protein 1; valosin-containing protein (p97)/p47 complex-interacting protein 135; valosin-containing protein (p97)/p47 complex-interacting protein p135; valosin-containing protein p97/p47 complex-interacting protein 1; Valosin-containing protein p97/p47 complex-interacting protein p135; VCIP135; VCP(p97)/p47-interacting protein; VCP/p47 complex-interacting 135-kDa protein; VCPIP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80124 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JH7 | |
VCPIP1 | |
Recombinant protein corresponding to Human VCPIP1. Recombinant protein control fragment (Product #RP-93910). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction