Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VCPKMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585003
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585003 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585003 Supplier Invitrogen™ Supplier No. PA585003
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EIEGFPSPPD FILMADCIYY EESLEPLLKT LKDISGFETC IICCYEQRTM GKNPEIEKKY FELLQLDFDF EKIPLEKHDE EYRSEDIHII YIRKK Highest antigen sequence indentity to the following orthologs: Rat - 27%, Mouse - 85%.

C14orf138, also named as VCPKMT or METTL21D, is 229 amino acid protein, which belongs to the methyltransferase superfamily. C14orf138 localizes in the cytoplasm and interacts with ALKBH6. C14orf138 as a Protein-lysine N-methyltransferase that specifically trimethylates 'Lys-315' of VCP/p97; this modification may decrease VCP ATPase activity. C14orf138 exists as five isoforms. The calculated molecular weight of C14orf138 is 26 kDa, but we always detected a 46-50 kDa protein by western blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VCPKMT
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VCPKMT
Gene Accession No. Q9H867
Gene Alias C14orf138; methyltransferase like 21D; methyltransferase-like protein 21D; METTL21D; Protein-lysine methyltransferase METTL21D; valosin containing protein lysine (K) methyltransferase; valosin containing protein lysine methyltransferase; valosin-containing protein lysine methyltransferase; VCP lysine methyltransferase; VCPKMT; VCP-KMT
Gene Symbols VCPKMT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VCPKMT. Recombinant protein control fragment (Product #RP-108662).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79609
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.