Learn More
Invitrogen™ VCX3A Polyclonal Antibody

Description
VCX3A Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | VCX3A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | VCX3A |
| Gene Accession No. | Q9NNX9 |
| Gene Alias | Variable charge protein on X with eight repeats; variable charge X-linked protein 3; variable charge, X-linked 3A; variably charged protein X-A; variably charged X-A; VCX3; VCX3A; VCX8R; VCX-8r; VCXA; VCX-A |
| Gene Symbols | VCX3A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.