Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VCY Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24670725UL
Description
VCY Polyclonal antibody specifically detects VCY in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
VCY | |
Polyclonal | |
Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
Basic Charge, Y-Linked 1, Basic Protein On Y Chromosome, 1, BPY1, BPY1B, Testis-Specific Basic Protein On Y, 1, Testis-Specific Basic Protein Y 1, Variable Charge, Y Chromosome, Variable Charge, Y-Linked, Variably Charged Protein Y, VCY VCY1B, VCY1, VCY1A | |
This antibody was developed against Recombinant Protein corresponding to amino acids: AATKMAAVTAPEAESGPAAPGPSDQPSQE | |
25 μL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9084 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction