Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VCY1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560888
Description
Immunogen sequence: AATKMAAVTA PEAESGPAAP GPSDQPSQE Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 48%.
The protein encoded by this gene is a member of a family of human VCX/Y genes. This gene family has multiple members on both X and Y chromosomes, and all are expressed exclusively in male germ cells. Members of the VCX/Y family share a high degree of sequence identity, with the exception that a 30-bp unit is tandemly repeated in X-linked members but occurs only once in Y-linked members. VCX/Y genes encode small and highly charged proteins of unknown function. This gene encodes a small, positively charged protein. The presence of a putative bipartite nuclear localization signal suggests that this gene encodes a nuclear protein. The genome has two identical copies of this gene within a palindromic region; this record represents the more centromeric copy.
Specifications
| VCY1B | |
| Polyclonal | |
| Unconjugated | |
| VCY | |
| Basic charge, Y-linked 1; basic protein on Y chromosome, 1; BPY1; BPY1B; testis-specific basic protein on Y, 1; testis-specific basic protein Y 1; variable charge, Y chromosome; variable charge, Y-linked; variably charged protein Y; VCY; VCY1; VCY1A; VCY1B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9084 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14598 | |
| VCY | |
| Recombinant protein corresponding to Human VCY1B. Recombinant protein control fragment (Product #RP-104377). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction