Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VDAC Monoclonal Antibody (C1), Invitrogen™

Catalog No. PIMA543951
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543951 20 μL
PIMA543950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543951 Supplier Invitrogen™ Supplier No. MA543951
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Voltage-dependent anion channel (VDAC) proteins are abundant, pore-forming proteins belonging to the eukaryotic mitochondrial porins. It was discovered in the mitochondrial outer membrane. It also is expressed in the plasma membrane. At least three different VDAC genes have been identified in vertebrates. VDAC proteins are known to play an essential role in cellular metabolism and in the early stages of apoptosis. For example, VDAC constitutes a major pathway by which metabolites such as ADP/ATP, succinate and citrate are exchanged between the cytosol and mitochondria. And VDAC1 in the plasma membrane establishes a novel level of apoptosis regulation putatively via its redox activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VDAC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene VDAC1
Gene Accession No. P21796
Gene Alias AL033343; BR1-VDAC; Brain-derived voltage-dependent anion channel 1; hVDAC1; MGC111064; Mitochondrial outer membrane protein porin 1; Mitochondrial Porin; mVDAC1; mVDAC5; N2441; OMP2; Outer mitochondrial membrane protein porin 1; plasmalemmal porin; POR1; PORIN; Porin 31HL; porin 31HM; porin POR1; PORIN-31-HL; rVDAC1; VD; VDAC; VDAC1; VDAC-1; Vdac5; VDAC-5; voltage dependent anion channel 1; voltage-dependent anion channel 1; voltage-dependent anion-selective channel protein 1; Voltage-dependent anion-selective channel protein 5; YNL055C; YNL2441C
Gene Symbols VDAC1
Host Species Mouse
Immunogen Ala2-Ala283 (Accession P21796), with N-terminal His and GST Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7416
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.