Learn More
Invitrogen™ VDAC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585405
Description
Keep as concentrated solution. Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (85%), Pig (98%), Rabbit (99%), Chicken (93%), Sheep (99%), Rhesus Monkey (98%), Bovine (99%). Positive Control: A431, H1299, HeLa, HepG2, F05, F32, F39, F90. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Voltage-dependent anion channel (VDAC) proteins are abundant, pore-forming proteins belonging to the eukaryotic mitochondrial porins. It was discovered in the mitochondrial outer membrane. It also is expressed in the plasma membrane. At least three different VDAC genes have been identified in vertebrates. VDAC proteins are known to play an essential role in cellular metabolism and in the early stages of apoptosis. For example, VDAC constitutes a major pathway by which metabolites such as ADP/ATP, succinate and citrate are exchanged between the cytosol and mitochondria. And VDAC1 in the plasma membrane establishes a novel level of apoptosis regulation putatively via its redox activity.
Specifications
VDAC | |
Polyclonal | |
Unconjugated | |
VDAC1 | |
AL033343; BR1-VDAC; Brain-derived voltage-dependent anion channel 1; hVDAC1; MGC111064; Mitochondrial outer membrane protein porin 1; Mitochondrial Porin; mVDAC1; mVDAC5; N2441; OMP2; Outer mitochondrial membrane protein porin 1; plasmalemmal porin; POR1; PORIN; Porin 31HL; porin 31HM; porin POR1; PORIN-31-HL; rVDAC1; VD; VDAC; VDAC1; VDAC-1; Vdac5; VDAC-5; voltage dependent anion channel 1; voltage-dependent anion channel 1; voltage-dependent anion-selective channel protein 1; Voltage-dependent anion-selective channel protein 5; YNL055C; YNL2441C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22333, 7416, 83529 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.46 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P21796, Q60932, Q9Z2L0 | |
VDAC1 | |
Recombinant protein encompassing a sequence within the center region of human VDAC1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.