Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VDAC/PorinRabbit anti-Bovine, Goat, Human, Mouse, Porcine, Rat, Polyclonal, Rockland™
SDP

Catalog No. 501991838
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1838 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1838 Supplier Rockland Immunochemicals Supplier No. 600401882RTU
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 2-8°C prior to opening. May aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. Dilute only prior to use. RTU Anti-VDAC/Porin Antibody is directed against human VDAC1/Porin1 protein. A BLAST analysis was used to suggest that this antibody would react with VDAC1/Porin1 from a wide range or organisms, including avian, mammalian, aquatic and reptilian sources based on 100% homology for the immunogen sequence. Cross reactivity will occur with all forms of VDACs including VDAC1, VDAC2 (4 isoforms) and VDAC3 (2 isoforms). Such broad reactivity makes this antibody useful as an excellent loading control (mitochondrial).

Voltage-dependent anion channel (VDAC) proteins are abundant, pore-forming proteins belonging to the eukaryotic mitochondrial porins. It was discovered in the mitochondrial outer membrane. It also is expressed in the plasma membrane. At least three different VDAC genes have been identified in vertebrates. VDAC proteins are known to play an essential role in cellular metabolism and in the early stages of apoptosis. For example, VDAC constitutes a major pathway by which metabolites such as ADP/ATP, succinate and citrate are exchanged between the cytosol and mitochondria. And VDAC1 in the plasma membrane establishes a novel level of apoptosis regulation putatively via its redox activity.

Specifications

Antigen VDAC/Porin
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.01% BSA, 0.15M NaCl, 25% glycerol and 0.01% sodium azide; pH 7.2
Gene VDAC1
Gene Accession No. P21796, P45879, Q60932, Q9MZ16, Q9Z2L0
Gene Alias AL033343; hVDAC1; MGC111064; Mitochondrial outer membrane protein porin 1; Mitochondrial Porin; mVDAC1; mVDAC5; N2441; OMP2; Outer mitochondrial membrane protein porin 1; Plasmalemmal porin; POR1; PORIN; Porin 31HL; Porin 31HM; porin POR1; PORIN-31-HL; rVDAC1; VD; VDAC; Vdac1; VDAC-1; Vdac5; VDAC-5; voltage dependent anion channel 1; voltage-dependent anion channel 1; Voltage-dependent anion-selective channel protein 1; Voltage-dependent anion-selective channel protein 5; YNL055C; YNL2441C
Gene Symbols VDAC1
Host Species Rabbit
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102173290, 22333, 282119, 397010, 7416, 83529
Target Species Bovine, Goat, Human, Mouse, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.