Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VDAC1 Antibody (S152B-23), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242187H
Description
VDAC1 Monoclonal specifically detects VDAC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
VDAC1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
VDAC1 | |
Fusion protein amino acids 1-283 (full-length) of human VDAC1. Mouse: 98% identity (279/283 amino acids identical). Rat: 98% identity (279/283 amino acids identical) >60% identity with VDAC2 and VDAC3. | |
0.1 mL | |
Primary | |
Greater then 60% identity with VDAC2 and VDAC3.Detects approx 30kDa. Does not cross-react with VDAC2 or VDAC3 (based on KO validation results). | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunohistochemistry | |
S152B-23 | |
Western Blot, Immunohistochemistry | |
hVDAC1, MGC111064, Outer mitochondrial membrane protein porin 1, Plasmalemmal porin, PORIN, Porin 31HL, Porin 31HM, PORIN-31-HL, VDAC, VDAC-1, voltage-dependent anion channel 1, voltage-dependent anion-selective channel protein 1 | |
Mouse | |
Protein G purified | |
Cellular Markers, Mitochondrial Markers | |
7416 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction