Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VDAC1 Monoclonal Antibody (9A7), Bioss
SDP

Catalog No. 502281502
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1502 Supplier Bioss Supplier No. BSM52251R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

VDAC1 Monoclonal antibody specifically detects VDAC1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen VDAC1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 9A7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide
Gene VDAC1
Gene Accession No. P21796, Q60932, Q9Z2L0
Gene Alias AL033343; BR1-VDAC; Brain-derived voltage-dependent anion channel 1; hVDAC1; MGC111064; Mitochondrial outer membrane protein porin 1; Mitochondrial Porin; mVDAC1; mVDAC5; N2441; OMP2; Outer mitochondrial membrane protein porin 1; plasmalemmal porin; POR1; PORIN; Porin 31HL; porin 31HM; porin POR1; PORIN-31-HL; rVDAC1; VD; VDAC; VDAC1; VDAC-1; Vdac5; VDAC-5; voltage dependent anion channel 1; voltage-dependent anion channel 1; voltage-dependent anion-selective channel protein 1; Voltage-dependent anion-selective channel protein 5; YNL055C; YNL2441C
Host Species Rabbit
Immunogen Human VDAC1 protein between 1 to 100 amino acids
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22333, 7416, 83529
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.