Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VDAC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116887 Supplier Invitrogen™ Supplier No. PA5116887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-251MG, HepG2, HeLa, Mouse testis, Mouse kidney, Mouse brain, Rat kidney, Rat heart, Rat brain Immunogen sequence: DIFNKGFGFG L.

This gene encodes a member of the voltage-dependent anion channel pore-forming family of proteins that are considered the main pathway for metabolite diffusion across the mitochondrial outer membrane. The encoded protein is also thought to be involved in the mitochondrial apoptotic pathway via regulation of BCL2-antagonist/killer 1 protein activity. Pseudogenes have been identified on chromosomes 1, 2, 12 and 21, and alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VDAC2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Vdac2
Gene Accession No. P45880, P81155, Q60930
Gene Alias B36-VDAC; FLJ23841; mVDAC2; mVDAC6; Outer mitochondrial membrane protein porin 2; POR; RP11-375G3.1; VDAC; Vdac2; VDAC-2; VDAC2 protein; Vdac6; VDAC-6; voltage dependent anion channel 2; voltage-dependent anion channel 2; voltage-dependent anion-selective channel protein 2; Voltage-dependent anion-selective channel protein 6
Gene Symbols Vdac2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human VDAC2 (NP_0033662).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22334, 7417, 83531
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.