Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VDAC2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518264
Description
This antibody is predicted to react with bovine, canine, mouse and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene encodes a member of the voltage-dependent anion channel pore-forming family of proteins that are considered the main pathway for metabolite diffusion across the mitochondrial outer membrane. The encoded protein is also thought to be involved in the mitochondrial apoptotic pathway via regulation of BCL2-antagonist/killer 1 protein activity. Pseudogenes have been identified on chromosomes 1, 2, 12 and 21, and alternative splicing results in multiple transcript variants.
Specifications
VDAC2 | |
Polyclonal | |
Unconjugated | |
Vdac2 | |
B36-VDAC; FLJ23841; mVDAC2; mVDAC6; Outer mitochondrial membrane protein porin 2; POR; RP11-375G3.1; VDAC; Vdac2; VDAC-2; VDAC2 protein; Vdac6; VDAC-6; voltage dependent anion channel 2; voltage-dependent anion channel 2; voltage-dependent anion-selective channel protein 2; Voltage-dependent anion-selective channel protein 6 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
7417, 83531 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P45880, P81155 | |
Vdac2 | |
Synthetic peptide sequence (GHKVGLALELEA) corresponding to the C-terminus amino acids of VDAC2 (aa 283-294). | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction