Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VDAC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116112
Description
Antibody detects endogenous levels of total VDAC2.
This gene encodes a member of the voltage-dependent anion channel pore-forming family of proteins that are considered the main pathway for metabolite diffusion across the mitochondrial outer membrane. The encoded protein is also thought to be involved in the mitochondrial apoptotic pathway via regulation of BCL2-antagonist/killer 1 protein activity. Pseudogenes have been identified on chromosomes 1, 2, 12 and 21, and alternative splicing results in multiple transcript variants.
Specifications
VDAC2 | |
Polyclonal | |
Unconjugated | |
Vdac2 | |
B36-VDAC; FLJ23841; mVDAC2; mVDAC6; Outer mitochondrial membrane protein porin 2; POR; RP11-375G3.1; VDAC; Vdac2; VDAC-2; VDAC2 protein; Vdac6; VDAC-6; voltage dependent anion channel 2; voltage-dependent anion channel 2; voltage-dependent anion-selective channel protein 2; Voltage-dependent anion-selective channel protein 6 | |
Rabbit | |
Affinity chromatography | |
RUO | |
22334, 7417, 83531 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P45880, P81155, Q60930 | |
Vdac2 | |
A synthesized peptide derived from human VDAC2(Accession P45880), corresponding to amino acid residues R263-A294. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction