Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VDAC2 Recombinant Rabbit Monoclonal Antibody (5R5V8)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551321
Description
Immunogen sequence: MATHGQTCAR PMCIPPSYAD LGKAARDIFN KGFGFGLVKL DVKTKSCSGV EFSTSGSSNT DTGKVTGTLE TKYKWCEYGL TFTEKWNTDN TLGTEIAIED.
This gene encodes a member of the voltage-dependent anion channel pore-forming family of proteins that are considered the main pathway for metabolite diffusion across the mitochondrial outer membrane. The encoded protein is also thought to be involved in the mitochondrial apoptotic pathway via regulation of BCL2-antagonist/killer 1 protein activity. Pseudogenes have been identified on chromosomes 1, 2, 12 and 21, and alternative splicing results in multiple transcript variants.
Specifications
VDAC2 | |
Recombinant Monoclonal | |
0.6 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P45880, P81155, Q60930 | |
Vdac2 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human VDAC2 (NP_003366.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
5R5V8 | |
Unconjugated | |
Vdac2 | |
B36-VDAC; FLJ23841; mVDAC2; mVDAC6; Outer mitochondrial membrane protein porin 2; POR; RP11-375G3.1; VDAC; Vdac2; VDAC-2; VDAC2 protein; Vdac6; VDAC-6; voltage dependent anion channel 2; voltage-dependent anion channel 2; voltage-dependent anion-selective channel protein 2; Voltage-dependent anion-selective channel protein 6 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22334, 7417, 83531 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction