Learn More
VDAC2/VDAC3, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA555444
Description
The Voltage-Dependant Anion Channel (VDAC) of the outer mitochondrial membrane, a small abundant protein found in all eukaryotic kingdoms, forms a voltage-gated pore when incorporated into planar lipid bilayers. VDAC is also the site of binding of the metabolic enzymes hexokinase and glycerol kinase to the mitochondrion in what may be a significant metabolic regulatory interaction. VDAC3 is involved specifically in translocation of adenine nucleotides through the outer membrane.
Immunogen sequence: TSGHAYTDTG KASGNLETKY KVCNYGLTFT QKWNTDNTLG TEISWENKLA EGLKLTLDTI FVPNTGKKSG KLKASYKRDC FSVGSNVDID FSGPTIYGWA VLAFEGWLAG YQMSFDTAKS KLSQNNFALG YKAADFQLHT HHighest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 96%.
Specifications
| VDAC2/VDAC3 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P45880, Q9Y277 | |
| VDAC2, VDAC3 | |
| Recombinant protein corresponding to Human VDAC2/VDAC3 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| Unconjugated | |
| VDAC3 | |
| VDAC2, VDAC3, RP11-375G3.1, FLJ23841, Voltage-dependent anion-selective channel protein 2, Voltage-dependent anion-selective channel protein 3, hVDAC-2, hVDAC-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7417, 7419 | |
| Store at 4°C short term. Store at −20°C long term. Avoid freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only