Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VDAC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100190 Supplier Invitrogen™ Supplier No. PA5100190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total VDAC3.

The Voltage-Dependent Anion Channel (VDAC) of the outer mitochondrial membrane, a small abundant protein found in all eukaryotic kingdoms, forms a voltage-gated pore when incorporated into planar lipid bilayers. VDAC is also the site of binding of the metabolic enzymes hexokinase and glycerol kinase to the mitochondrion in what may be a significant metabolic regulatory interaction. VDAC3 is involved specifically in translocation of adenine nucleotides through the outer membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VDAC3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene VDAC3
Gene Accession No. Q60931, Q9R1Z0, Q9Y277
Gene Alias channel protein VDAC3; HD-VDAC3; mitochondrial outer membrane protein; mitochondrial voltage dependent anion channel 3; mVDAC3; outer mitochondrial membrane protein porin 3; rVDAC3; Vdac3; VDAC-3; voltage dependent anion channel 3; voltage-dependent anion channel 3; voltage-dependent anion-selective channel protein 3
Gene Symbols VDAC3
Host Species Rabbit
Immunogen A synthesized peptide derived from human VDAC3(Accession Q9Y277), corresponding to amino acid residues D30-T80.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22335, 7419, 83532
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.