Learn More
Invitrogen™ VDAC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587973
Description
Positive Samples: A-431; Cellular Location: Mitochondrion outer membrane Immunogen sequence: MCNTPTYCDL GKAAKDVFNK GYGFGMVKID LKTKSCSGVE FSTSGHAYTD TGKASGNLET KYKVCNYGLT FTQKWNTDNT LGTEISWENK LAEGLKLTLD TIFVPNTGKK SGKLKASYKR DCFSVGSNVD IDFSGPTIYG WAVLAFEGWL AGYQMSFDTA KSKLSQNNFA LGYKAADFQL HTHVNDGTEF GGSIYQKVNE KIETSINLAW TAGSNNTRFG IAAKYMLDCR TSLSAKVNNA SLIGLGYTQT LRPGVKLTLS ALIDGKNFSA GGHKVGLGFE LEA.
The Voltage-Dependent Anion Channel (VDAC) of the outer mitochondrial membrane, a small abundant protein found in all eukaryotic kingdoms, forms a voltage-gated pore when incorporated into planar lipid bilayers. VDAC is also the site of binding of the metabolic enzymes hexokinase and glycerol kinase to the mitochondrion in what may be a significant metabolic regulatory interaction. VDAC3 is involved specifically in translocation of adenine nucleotides through the outer membrane.
Specifications
VDAC3 | |
Polyclonal | |
Unconjugated | |
VDAC3 | |
channel protein VDAC3; HD-VDAC3; mitochondrial outer membrane protein; mitochondrial voltage dependent anion channel 3; mVDAC3; outer mitochondrial membrane protein porin 3; rVDAC3; Vdac3; VDAC-3; voltage dependent anion channel 3; voltage-dependent anion channel 3; voltage-dependent anion-selective channel protein 3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22335, 7419 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
3.93 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q60931, Q9Y277 | |
VDAC3 | |
A synthetic peptide corresponding to a sequence within amino acids 1-283 of human VDAC3 (NP_0056533). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.