Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VDP p115 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074483
Description
VDP p115 Polyclonal specifically detects VDP p115 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
VDP p115 | |
Polyclonal | |
Unconjugated | |
PBS with 1mg/ml BSA with 0.05% Sodium Azide | |
p115, Transcytosis-associated protein, USO1 homolog, vesicle docking protein (yeast), USO1 vesicle docking protein homolog (yeast), vesicle docking protein, vesicle docking protein p115 | |
Rabbit | |
Affinity Purified | |
RUO | |
8615 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 1:500, Flow Cytometry 3 - 5 ug/10^6 cells, Immunocytochemistry/Immunofluorescence 2 - 3 ug/mL | |
P41542 | |
USO1 | |
Synthetic peptide corresponding to residues M(1) N F L R G V M G G Q S A G P Q H(17) of rat p115. | |
100 μL | |
Primary | |
Rat VDP p115. The sequence is conserved across many species including mouse and human. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction