Learn More
Invitrogen™ VDR Polyclonal Antibody

Description
This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
Specifications
Specifications
| Antigen | VDR |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | VDR |
| Gene Accession No. | P11473 |
| Gene Alias | 1,25-dihydroxyvitamin D3 receptor; HGNC:12679; I79_025795; Nr1i1; Nuclear receptor subfamily 1 group I member 1; PPP1R163; protein phosphatase 1, regulatory subunit 163; vdr; vdr.S; vdr-A; vitamin D (1,25- dihydroxyvitamin D3) receptor; vitamin D (1,25- dihydroxyvitamin D3) receptor S homeolog; vitamin D (1,25-dihydroxyvitamin D3) receptor; vitamin D nuclear receptor variant 1; vitamin D receptor; vitamin D receptor protein; vitamin D receptor S homeolog; Vitamin D3 receptor; XELAEV_18015993mg; xVDR |
| Gene Symbols | VDR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.