Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VE-Cadherin Antibody (16B1), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200342AF532
Description
VE-Cadherin Monoclonal specifically detects VE-Cadherin in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
VE-Cadherin | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CDH5 | |
The immunogen for this antibody is VE Cadherin. | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry | |
16B1 | |
Western Blot, Flow Cytometry | |
7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin | |
Mouse | |
0.1 mL | |
1003 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction