Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VE-Cadherin Antibody (201), Alexa Fluor™ 647

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290471AF647
Description
VE-Cadherin Monoclonal specifically detects VE-Cadherin in Mouse samples. It is validated for Flow Cytometry.Specifications
VE-Cadherin | |
Monoclonal | |
Alexa Fluor 647 | |
7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse VE-Cadherin (NP_033998.2; Met1-Ala592). | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
1003 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry | |
201 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction